Vane spindles for viscometers
: AMETEK
Vane spindles can be used with any standard viscometers.
V-74 vane spindle is suitable for testing small samples and fitting into sample containers with small openings.
Frequently Bought Together
Vane spindles can be used with any standard viscometers.
V-74 vane spindle is suitable for testing small samples and fitting into sample containers with small openings.