Order Entry
Italy
ContactUsLinkComponent
Hydrophobic interaction chromatography column, HiScreen™ Capto™ Phenyl ImpRes
Hydrophobic interaction chromatography column, HiScreen™ Capto™ Phenyl ImpRes
Catalog # 17-5484-10
Supplier:  Cytiva
Hydrophobic interaction chromatography column, HiScreen™ Capto™ Phenyl ImpRes
Catalog # 17-5484-10
Supplier:  Cytiva

Specifications

  • Capacity:
    4,7 ml
  • Description:
    HiScreen™ Capto™ Phenyl ImpRes
  • Bed dimensions:
    7,7×100 mm
  • Pk:
    4,7 mL

Specifications

About this item

HiScreen™ Capto Phenyl ImpRes columns are prepacked with Capto™ Phenyl ImpRes hydrophobic interaction chromatography medium. The columns are an excellent choice for method optimisation and parameter screening.

  • Easily connected in series to achieve 20 cm bed height
  • Small bed volume gives fast results and minimal sample/buffer consumption

Reproducible results, scalable to BioProcess columns packed with the same chromatography resins using the same linear fluid velocity.