Order Entry
Italy
ContactUsLinkComponent
Hydrophobic interaction chromatography column, HiScreen™ Capto™ Butyl ImpRes
Hydrophobic interaction chromatography column, HiScreen™ Capto™ Butyl ImpRes
Catalog # 17-3719-10
Supplier:  Cytiva
Hydrophobic interaction chromatography column, HiScreen™ Capto™ Butyl ImpRes
Catalog # 17-3719-10
Supplier:  Cytiva

Specifications

  • Max. pressure:
    3 bar
  • Capacity:
    4,7 ml
  • Description:
    HiScreen™ Capto™ Butyl ImpRes
  • Storage:
    4 to 30 °C, 20% Ethanol
  • Bed dimensions:
    7,7×100 mm
  • Pk:
    1

Specifications

About this item

HiScreen™ Capto™ Butyl ImpRes is a ready-to-use column prepacked with Capto™ Butyl ImpRes hydrophobic interaction chromatography resin.

  • Excellent for method optimisation and parameter screening
  • Easily connected in series to achieve 20 cm bed height
  • Small bed volume gives fast results and minimal sample/buffer consumption

Reproducible results, scalable to BioProcess columns packed with the same chromatography resin using the same linear fluid velocity.