Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Colorimeters
Comparator Kits
Accessories for Nessleriser systems
Accessories for Nessleriser systems
Accessories for Nessleriser systems
Catalog # TINT354230
Supplier:
Lovibond Tintometer
Expand image
Accessories for Nessleriser systems
Catalog # TINT354230
Supplier:
Lovibond Tintometer
Specifications
Description:
DB423 pair of Nessler tubes, 100 ml/288 mm (ASTM D 1209) with glass anti-meniscus plungers
Pk:
2
Specifications
About this item
Product Details
Product Details