Order Entry
Italy
ContactUsLinkComponent
4,4'-Bipyridyl for synthesis, Sigma-Aldrich®
Toxic
4,4'-Bipyridyl for synthesis, Sigma-Aldrich®
Catalog # 8.20158.0010
Supplier:  Merck
CAS Number:  
Toxic
4,4'-Bipyridyl for synthesis, Sigma-Aldrich®
Catalog # 8.20158.0010
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Solubility:
    4.5 g/l (25 °C)
  • Melting Point:
    109 - 113 °C
  • Molar Mass:
    156.18 g/mol
  • Boiling Point:
    305 °C

Specification Test Results

About this item

 :
 :156.19 g/mol
Merck Index:13,03383
Boiling Pt:305 °C (1013 hPa)
Melting Pt:78 °C
Storage Temperature:Ambient
EINECS:209-036-3
UN:2811
ADR:6.1,II
 :M-Clarity™ quality level = MQ200
 :GHS06
CAS Number:553-26-4
Formula:C₁₀H₈N₂
MDL Number:MFCD00006416