
6-Methyl-2-pyridone ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3279-76-3
:
Formula:C₆H₇NO
MDL Number:MFCD00006279
Boiling Pt:283…284 °C (1013 hPa)
Melting Pt:157…161 °C
Density:1.13 g/cm³ (20 °C)
:109.13 g/mol
Storage Temperature:Ambient
EINECS:221-919-5
:GHS07
Related Information
Frequently Bought Together