Order Entry
Italy
ContactUsLinkComponent
Sterile vial kits, WHEATON® CompletePAK™
Sterile vial kits, WHEATON® CompletePAK™
Catalog # 218-0047
Supplier:  DWK Life Sciences
Sterile vial kits, WHEATON® CompletePAK™
Catalog # 218-0047
Supplier:  DWK Life Sciences

Specifications

  • Cap included:
    No
  • Cap Size:
    20 mm
  • Capacity:
    5,00 ml
  • Colour:
    Amber
  • Description:
    Sterile serum vial
  • Seq:
    43
  • Pk:
    200

Specifications

About this item

CompletePAK™ is a range of primary packaging components that are stocked ready to use and are customisable for most drug applications.

  • Off the shelf, ready to use products
  • One source for total packaging solutions
  • Variety of different sterile components; a kit for any application
  • Complete manufacturing traceability

Each kit component is particulate cleaned, depyrogenated, and sterilised, arriving in the lab ready to use. Vials are packed in a sterile PP tray which can be used as a filling aid. Kit packaging is sized to compliment manual aseptic filling processes.

The CompletePAK™ 'single source' focus can reduce customer's supply chain and time to market.

Certifications: Certificates of Analysis in each kit shows compliance with:
1. USP <788>/EP 2.9.19 Particulate Matter in Injections
2. USP <85>/EP 2.6.1 The Bacterial Endotoxins Test
3. USP <71>/ EP 2.6.14 The Sterility Test
4. USP <660>/EP 3.2.1 Glass Containers for Pharmaceutical Use
5. USP <381>/EP 3.2.9 Elastomeric Closures used in Parenteral Preparations
Delivery: Batch-specific Certificates of Analysis showing compliance with relevant USP/EP standards are included in each kit.

CompletePAK® sterile stoppers only

Product Information