
5-Bromo-2-methoxypyridine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13472-85-0
:
Formula:C₆H₆BrNO
MDL Number:MFCD01318952
Boiling Pt:80 °C (12 mmHg)
Flash Pt:96 °C
:188.02 g/mol
:GHS07
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5530 to 1.5570 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together