
5-Bromonicotinic acid 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:20826-04-4
:
Formula:C₆H₄BrNO₂
MDL Number:MFCD00009783
:202.01 g/mol
Boiling Pt:300 °C (1013 hPa)
Melting Pt:178…180 °C
Storage Temperature:Ambient
EINECS:244-065-5
:GHS07
Specification Test Results
- Appearance:White to yellow-grey to light brown Powder
- Infrared spectrum:Conforms
- Melting point:≥180 °C
- HPLC:≥97.5 %
- Titration with TBAH:≥97.5 %
Specification Test Results
Frequently Bought Together