Order Entry
Italy
ContactUsLinkComponent
Dimidium bromide, GPR RECTAPUR®
Irritant
Dimidium bromide, GPR RECTAPUR®
Catalog # 201272P
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Dimidium bromide, GPR RECTAPUR®
Catalog # 201272P
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass ampoule
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Dye content (spectrophotometrically):
    Min. 95 %
  • Identity (UV/VIS-Spectrum):
    Passes test
  • Absorption max. (in methanol):
    523 - 528 nm
  • Spec.Absorptivity (?m;0,0005 %;methanol):
    166 - 175
  • TLC-Test:
    Passes test
  • Loss on drying (110°C):
    Max. 5 %

Specification Test Results

About this item

 :
 :380.29 g/mol
Melting Pt:243…248 °C
Storage Temperature:Ambient
EINECS:208-256-7
UN:2811
ADR:6.1,I
 :GHS07
CAS Number:518-67-2
Formula:C₂₀H₁₈BrN₃
MDL Number:MFCD00011757