3,6-Dimethylphenanthrene
Catalog # EHERC20785000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:206.29 g/mol
EINECS:216-409-4
CAS Number:1576-67-6
Formula:C₁₆H₁₄
MDL Number:MFCD00001170