Order Entry
Italy
ContactUsLinkComponent
Phenanthrene for synthesis, Sigma-Aldrich®
Irritant
Environment
Phenanthrene for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
undefined
Irritant
Environment
Phenanthrene for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    1.15 mg/l (25 °C)
  • Melting Point:
    97 - 100 °C
  • Molar Mass:
    178.23 g/mol
  • Bulk Density:
    690 kg/m3
  • Boiling Point:
    336 °C (1013 hPa)
  • Vapor Pressure:
    0.00012 hPa (20 °C)
  • Flash Point:
    171 °C
  • Ignition Point:
    450 °C

Specification Test Results

Related Information

Frequently Bought Together