Order Entry
Italy
ContactUsLinkComponent
 
Human Recombinant GDF-15-D MUTANT (from E. coli)
 
undefined
Human Recombinant GDF-15-D MUTANT (from E. coli)

 

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    −20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤1
  • Biological activity:
    Bioactivity assay: No test available, ED50 ≤Currently undetermined, bioactivity ≥Currently undetermined
  • Gene ID:
    Q99988
  • Reconstitution instructions:
    Sterile water at 0,1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    MIC-1|Prostate differentiation factor|Placental TGF-β
  • UniProtKB:
    Q99988
  • Protein/peptide name:
    GDF-15 D
  • Purity:
    ≥95%
  • Molecular weight:
    12,4/24,8 kDa
  • Sequence:
    MARNGDDCPL GPGRCCRLHT VRASLEDLGW ADWVLSPREV QVTMCIGACP SQFRAANMHA QIKTSLHRLK PDTVPAPCCV PASYNPMVLI QKTDTGVSLQ TYDDLLAKDC HCI
  • Endotoxin level:
    Low
  • Formulation:
    0,1% Trifluoroacetic Acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT

 

Frequently Bought Together