About this item
Synonyms: Sorbitan monododecanoate, Arlacel 20, Sorbitan monolaurate
- CAS Number:1338-39-2
- Formula:C₁₈H₃₄O₆
- MDL Number:MFCD00005365
- Molecular Weight:346.46 g/mol
- Merck Index:12,08872
- Density:1.05 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ200
- Storage Temperature:Ambient
- EINECS:215-663-3
3 Options Available Below
Product Details & Documents
Synonyms: Sorbitan monododecanoate, Arlacel 20, Sorbitan monolaurate
- CAS Number:1338-39-2
- Formula:C₁₈H₃₄O₆
- MDL Number:MFCD00005365
- Molecular Weight:346.46 g/mol
- Merck Index:12,08872
- Density:1.05 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ200
- Storage Temperature:Ambient
- EINECS:215-663-3
Documents
Certificates
Search by Lot Numbermsds_840119
SDS
ProductPage_840119 ProductPage_840119
Product specification
Specifications for Product Family
Specification Test Results
- Molar Mass:346.47 g/mol
- Boiling Point:100 °C (1013 hPa)
- Vapor Pressure:- 1.3 hPa (20 °C)
- Flash Point:149 °C
- Density:1.0 g/cm3 (20 °C)



