Succinic anhydride 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:108-30-5
:
Formula:C₄H₄O₃
MDL Number:MFCD00005525
:100.07 g/mol
Merck Index:12,09039
Boiling Pt:261 °C (760 mmHg)
Melting Pt:116…120 °C
Density:1,503 g/cm³ (20 °C)
Flash Pt:157 °C
Storage Temperature:Ambient
EINECS:203-570-0
UN:3261
ADR:8,II
:GHS05
:GHS07
:GHS08
Specification Test Results
- Appearance:White Powder or crystals or flakes and/or chips
- Infrared spectrum:Conforms
- Melting point:116 to 120 °C
- Titration by coupling:≥98.5 %
Specification Test Results
Frequently Bought Together