Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Kojic acid 99%

Health
Kojic acid 99%
Certificates
Kojic acid 99%

4 Options Available Below

Product Details & Documents
Synonyms:  5-Hydroxy-2-hydroxymethyl-4H-pyran-4-one, 5-Hydroxy-2-(hydroxymethyl)-4H-pyran-4-one, 5-Hydroxy-2-hydroxymethyl-4-pyrone, 5-Hydroxy-2-hydroxymethyl-γ-pyrone
  • CAS Number:501-30-4
  • Formula:C₆H₆O₄
  • MDL Number:MFCD00006580
  • Molecular Weight:142.11 g/mol
  • Merck Index:13,05334
  • Boiling Pt:323 °C (1013 hPa)
  • Melting Pt:151…156 °C
  • Density:1,58 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:207-922-4

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to beige to light brown Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    151 to 156 °C
  • Titration with NaOH:
    ≥98.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 45,60 €
    45,60 €
    1 * 5 g
       45,60 €

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 139,00 €
    139,00 €
    1 * 25 g
       139,00 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 495,00 €
    495,00 €
    1 * 100 g
       495,00 €

  • Glass bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 1 430,00 €
    1 430,00 €
    1 * 500 g
       1 430,00 €