


Cobalt(II) sulphate heptahydrate 97.0-102.0% (by complexometric analysis), extra pure, SAFC®
Catalog # 1.02546.9050
Supplier: MERCK PRODUCTION CHEMICALS
CAS Number:
Specifications
- Pack type:Cardboard carton
- Pk:50 kg
- Seq:4418
Specifications
Specification Test Results
- Assay (complexometric):97.0 - 102.0 %
- Identity:conforms
- Chloride (Cl):≤ 0.02 %
- Fe (Iron):≤ 0.005 %
- Pb (Lead):≤ 0.005 %
- Residual solvents (according to ICH-guideline):excluded by manufacturing process
Specification Test Results
About this item
:
:281.1 g/mol
Merck Index:12,02510
Melting Pt:735 °C
Density:2.03 g/cm³ (20 °C)
EINECS:233-334-2
UN:3077
ADR:9,III
:GHS08
:GHS07
:GHS09
CAS Number:10026-24-1
Formula:CoSO₄·7H₂O
MDL Number:MFCD00149658