

Nickel (II) oxide ≥99% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1313-99-1
:
Formula:NiO
MDL Number:MFCD00011145
:74.69 g/mol
Merck Index:14,6512
Melting Pt:1984 °C
Density:6.67
EINECS:215-215-7
:GHS08
:GHS07
Storage Temperature:Ambient
Frequently Bought Together