

Cobalt(II) oxide ≥99.995% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1307-96-6
:
Formula:CoO
MDL Number:MFCD00016031
:74.93 g/mol
Merck Index:14,2446
Melting Pt:1935 °C
Density:6.45
Storage Temperature:Ambient
EINECS:215-154-6
UN:2811
ADR:6.1,III
:GHS08
:GHS07
Related Information
Frequently Bought Together