Order Entry
Italy
ContactUsLinkComponent
Platinum(IV) chloride ≥99.99% (metals basis) Pt ≥ 57%, Premion®
Toxic
Health
Corrosive
Platinum(IV) chloride ≥99.99% (metals basis) Pt ≥ 57%, Premion®
Catalog # 44503.06
CAS Number:  
Toxic
Health
Corrosive
Platinum(IV) chloride ≥99.99% (metals basis) Pt ≥ 57%, Premion®
Catalog # 44503.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

About this item

CAS Number:13454-96-1
 :
Formula:Cl₄Pt
MDL Number:MFCD00011182
Melting Pt:370 °C (dec.)
Density:4.303
Storage Temperature:Ambient
EINECS:236-645-1
UN:2923
ADR:8,III
 :336.89 g/mol
 :GHS06
 :GHS08
 :GHS05