
Platinum(II) chloride (≥73% Pt) for synthesis, Sigma-Aldrich®
Catalog # 8.24566.0500
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mg
- Seq:2725
Specifications
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:581 °C decomposes
- Molar Mass:265.99 g/mol
- Density:6.1 g/cm3
- pH:5 - 6 (H2O) suspension
Specification Test Results
About this item
:
:265.99 g/mol
Merck Index:12,07687
Melting Pt:581 °C
Density:6.05 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-034-1
UN:1759
ADR:8,III
:M-Clarity™ quality level = MQ200
:GHS08
CAS Number:10025-65-7
Formula:Cl₂Pt
MDL Number:MFCD00011181