

Platinum(II) chloride ≥99.99% (metals basis) Pt ≥ 73%, Premion®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10025-65-7
:
Formula:Cl₂Pt
MDL Number:MFCD00011181
:265.99 g/mol
Merck Index:14,7528
Melting Pt:581 °C (dec.)
Density:5.87
Storage Temperature:Ambient
EINECS:233-034-1
UN:3260
ADR:8,III
:GHS08
:GHS05
Related Information
Frequently Bought Together