Order Entry
Italy
ContactUsLinkComponent
Manganese(II) chloride for synthesis, Sigma-Aldrich®
Toxic
Corrosive
Health
Environment
Manganese(II) chloride for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Toxic
Corrosive
Health
Environment
Manganese(II) chloride for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    723 g/l (25 °C)
  • Melting Point:
    650 °C
  • Molar Mass:
    125.84 g/mol
  • Boiling Point:
    1190 °C
  • Density:
    2.977 g/cm3 (25 °C)
  • pH:
    5.5 (25 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Frequently Bought Together