

Tungsten(VI) chloride 99.9+% (trace metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13283-01-7
:
Formula:Cl₆W
MDL Number:MFCD00011463
Boiling Pt:347 °C (760 mmHg)
Melting Pt:275 °C
Density:3,518 g/cm³ (20 °C)
EINECS:236-293-9
UN:3260
ADR:8,II
:396.56 g/mol
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Grey to dark blue to violet to black Powder
- Total trace metal impurities:≤0.10 %
Specification Test Results
Frequently Bought Together