


Chromium(III) chloride, anhydrous 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10025-73-7
:
Formula:Cl₃Cr
MDL Number:MFCD00010948
:158.35 g/mol
Merck Index:14,2222
Boiling Pt:1300 °C (subl.)
Melting Pt:1152 °C
Density:2.87
Storage Temperature:Ambient
EINECS:233-038-3
UN:3260
ADR:8,III
:GHS08
:GHS05
:GHS07
Related Information
Frequently Bought Together