Order Entry
Italy
ContactUsLinkComponent
Niobium(V) oxide ≥99.95% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Niobium(V) oxide ≥99.95% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Catalog # 41080.KS
CAS Number:  
Niobium(V) oxide ≥99.95% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Catalog # 41080.KS
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

CAS Number:1313-96-8
 :
Formula:Nb₂O₅
MDL Number:MFCD00011128
 :265.81 g/mol
Merck Index:14,06559
Melting Pt:1496 °C
Density:4.6 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-213-6