Order Entry
Italy
ContactUsLinkComponent
Cadmium(II) carbonate ≥99.99% (metals basis)
Irritant
Environment
Cadmium(II) carbonate ≥99.99% (metals basis)
Catalog # 11864.14
CAS Number:  
Irritant
Environment
Cadmium(II) carbonate ≥99.99% (metals basis)
Catalog # 11864.14
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :172.42 g/mol
Merck Index:14,1616
Melting Pt:500 °C (dec.)
Density:4.26
Storage Temperature:Ambient
EINECS:208-168-9
UN:2570
ADR:6.1,III
 :GHS07
 :GHS09
CAS Number:513-78-0
Formula:CCdO₃
MDL Number:MFCD00010918