Order Entry
Italy
ContactUsLinkComponent
Silver carbonate 99%, pure
Corrosive
Environment
Silver carbonate 99%, pure
Certificates
undefined
Corrosive
Environment
Silver carbonate 99%, pure

Specification Test Results

  • Appearance:
    Yellow to green Granular powder
  • Assay:
    ≥98.5 %
  • Melting point:
    176 to 180 °C
  • Water:
    ≤1 %
  • HPLC:
    ≥97.5 %
  • Specific optical rotation:
    +10° to +12° (20 °C, 589 nm) (5 % in water)

Specification Test Results

Related Information

Frequently Bought Together