Order Entry
Italy
ContactUsLinkComponent
Copper(I) chloride 97+%, extra pure
Corrosive
Irritant
Environment
Copper(I) chloride 97+%, extra pure
Certificates
undefined
Corrosive
Irritant
Environment
Copper(I) chloride 97+%, extra pure
Specification Test Results
  • Appearance:
    Light green to grey Crystalline powder
  • Titration Complexometric:
    97.0 to 103.0 %
  • Sulfate (SO4):
    ≤500 ppm
  • Arsenic (As):
    ≤5 ppm
  • Iron (Fe):
    ≤100 ppm
  • Insoluble matter:
    ≤0.05 % (in acid)
  • Subst nt pptd by H2S(SO4):
    ≤0.5 %