Supelco® Organic Reference Standard, 1-Methylnaphthalene
Catalog # 45795-250MG
Supplier: Merck
CAS Number:
Specifications
- Pk:250 mg
- Standards:Analytical standard
- CAS No.:90-12-0
- Description:1-Methylnaphthalene
- Storage conditions:2...30 °C
- Hazard class description:H302 - H304 - H411
- Shipping conditions:STD
- MDL No.:MFCD00004034
- M-Clarity quality level:MQ100
- Melting point:–22 °C (lit.)
- Formula:C₁₀H₇CH₃
- Molecular weight:142,20
- Density:1,001 g/ml at 25 °C (lit.)
- Flash point:82 °C - Closed cup
- Boiling point:240 - 243 °C (lit.)
Specifications
About this item
- GHS signal: GHS07, GHS09
Caution: P264 - P273 - P301 + P310 - P331 - P391 - P405