Order Entry
Italy
ContactUsLinkComponent
2,3-Dihydroxynaphthalene for synthesis, Sigma-Aldrich®
Corrosive
Irritant
2,3-Dihydroxynaphthalene for synthesis, Sigma-Aldrich®
Catalog # 8.14578.0050
Supplier:  Merck
CAS Number:  
Corrosive
Irritant
2,3-Dihydroxynaphthalene for synthesis, Sigma-Aldrich®
Catalog # 8.14578.0050
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 g
  • Seq:
    3368

Specifications

Specification Test Results

  • Solubility:
    4 g/l (35 °C)
  • Melting Point:
    161 - 164 °C
  • Molar Mass:
    160.17 g/mol
  • Bulk Density:
    500 kg/m3
  • Vapor Pressure:
    0.8 hPa (149 °C)
  • Flash Point:
    175 °C
  • pH:
    6 (1 g/l, H2O, 20 °C)
  • Ignition Point:
    330 °C

Specification Test Results

About this item

 :
 :160.17 g/mol
Boiling Pt:354 °C (1013 hPa)
Melting Pt:151…152 °C
Density:1.12 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
EINECS:202-156-7
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:92-44-4
Formula:C₁₀H₈O₂
MDL Number:MFCD00004073