
Terbium(III) fluoride, anhydrous ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13708-63-9
:
Formula:TbF₃
MDL Number:MFCD00049572
:215.92 g/mol
Boiling Pt:2277 °C (1013 hPa)
Melting Pt:1172 °C
Density:7.23 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:237-247-0
UN:3288
ADR:6.1,III
:GHS07
Related Information
Frequently Bought Together