
Cerium trifluoride, anhydrous ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7758-88-5
:
Formula:CeF₃
MDL Number:MFCD00010932
:197.11 g/mol
Merck Index:14,1998
Boiling Pt:∼2300 °C
Melting Pt:1460 °C
Density:6.16
EINECS:231-841-3
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together