Lutetium trichloride, anhydrous ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10099-66-8
:
Formula:Cl₃Lu
MDL Number:MFCD00011099
:281.33 g/mol
Merck Index:14,5615
Melting Pt:905 °C (subl.)
Density:3.98
Storage Temperature:Ambient
EINECS:233-240-1
Frequently Bought Together