
Gadolinium trifluoride, anhydrous ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13765-26-9
:
Formula:F₃Gd
MDL Number:MFCD00016113
:214.25 g/mol
Melting Pt:1231 °C
Density:7.1
Storage Temperature:Ambient
EINECS:237-369-4
UN:3288
ADR:6.1,III
:GHS07
Related Information
Frequently Bought Together