Order Entry
Italy
ContactUsLinkComponent
VWR®, Accessories for VWR® Semi-micro, Analytical or Precision Balances
VWR®, Accessories for VWR® Semi-micro, Analytical or Precision Balances
Catalog # 611-3864
Supplier:  VWR Collection
undefined
VWR®, Accessories for VWR® Semi-micro, Analytical or Precision Balances
Catalog # 611-3864
Supplier:  VWR Collection
Specifications
  • Description:
    Ioniser panel, allows user to retrofit an integrated ioniser
  • Seq:
    311
  • Pk:
    1
  • For:
    VWR semi-micro, analytical or precision balances fitted with draftshields which have 3 sliding doors