Order Entry
Italy
Orders LinkContactUsLinkComponent
Multiport connector caps, GL 25, DURAN®
Supplier:   DWK Life Sciences
Multiport connector caps, GL 25, DURAN®
Supplier:  DWK Life Sciences
Certificates
Multiport connector caps, GL 25, DURAN®
Multiport connector caps, GL 25, DURAN®
3 Options Available Below
Product Details & Documents
DURAN® GL 25 connector caps with multiple ports are suitable for a range of applications were small volumes of liquid need to be sampled or dispensed.
  • High temperature resistance (up to 180 °C)
  • Caps are fully autoclavable and reusable
The barbed male tubing connectors are made from a surgical grade stainless steel (316L) and will fit tubing with a Ø 3,2 mm.
Product Family Options
Product Information
  • For
    Availability
    Price
  • SCOT292601301
    2-port connector with cap
    2-port connector with cap
    GL 25 threaded bottles
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 81,60 €
    81,60 €
    1 * 1 pezzi
       81,60 €
  • SCOT292611302
    3-port connector with cap
    3-port connector with cap
    GL 25 threaded bottles
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 84,40 €
    84,40 €
    1 * 1 pezzi
       84,40 €
  • SCOT292621303
    4-port connector with cap
    4-port connector with cap
    GL 25 threaded bottles
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 87,10 €
    87,10 €
    1 * 1 pezzi
       87,10 €