0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Italy
ContactUsLinkComponent
Ninhydrin
Ninhydrin ACS
Ninhydrin ACS
:
Thermo Fisher Scientific
Ninhydrin ACS
:
Thermo Fisher Scientific
:
485-47-2
:
:
C₉H₆O₄
:
MFCD00003791
:
178.14 g/mol
:
14,6554
:
351 °C (1013 hPa)
:
251 °C (dec.)
:
0.862 g/cm³ (20 °C)
:
207-618-1
:
GHS07
:
Ambient
externalurl.pdf
msds_ALFA043846
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
: Merck
Sealing film for manual application, Parafilm® M
: HEATHROW
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals