
2,4-Dimethylimidazole ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:930-62-1
:
Formula:C₅H₈N₂
MDL Number:MFCD00022365
:96.13 g/mol
Boiling Pt:266 °C (733 mmHg)
Melting Pt:84…88 °C
Storage Temperature:Ambient
EINECS:213-221-4
UN:3263
ADR:8,III
:GHS05
Related Information
Frequently Bought Together