

1-Methylimidazole (NMI) 99%
:616-47-7
:
:C₄H₆N₂
:MFCD00005292
:198 °C (1013 hPa)
:–2 °C
:1.03 g/cm³
:92 °C
:Ambient
:210-484-7
:8,II
:82.11 g/mol
:2922
:GHS06
:GHS05
- Appearance:Clear colorless to yellow Liquid (May discolor to yellow on storage)
- Infrared spectrum:Conforms
- Refractive index:1.4955 to 1.4975 (20°C, 589 nm)
- Water:≤0.5 % (K.F.)
- GC:≥98.5 %
- Color scale:≤15 APHA
Frequently Bought Together