Imidazole ≥99.9%
Catalog # EHERC14283950
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
Merck Index:13,04935
Boiling Pt:255 °C (1013 hPa)
Melting Pt:89…90 °C
Density:1.18 g/cm³ (20 °C)
Flash Pt:135 °C
Storage Temperature:Ambient
EINECS:206-019-2
UN:2923
ADR:8,III
:68.08 g/mol
:GHS05
:GHS08
:GHS07
CAS Number:288-32-4
Formula:C₃H₄N₂
MDL Number:MFCD00005183