Imidazole 99+%, crystalline
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:288-32-4
:
Formula:C₃H₄N₂
MDL Number:MFCD00005183
Merck Index:13,04935
Boiling Pt:255…256 °C (760 mmHg)
Melting Pt:86…90 °C
Density:1,18 g/cm³ (20 °C)
Flash Pt:145 °C
Storage Temperature:Ambient
UN:3263
ADR:8,III
:68.08 g/mol
EINECS:206-019-2
:GHS05
:GHS07
:GHS08
Specification Test Results
- Appearance:White to light yellow to light beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:86 to 90 °C
- Water:≤1 % (K.F.)
- GC:≥99.0 %
- Titration with HClO4:≥99.0 % (On dry substance)
Specification Test Results
Related Information
Frequently Bought Together