Order Entry
Italy
ContactUsLinkComponent
 
 
 
Iodine monobromide 98%
 
undefined
 
 
Iodine monobromide 98%
 

 

  • Appearance:
    Dark grey to brown-black Crystals or crystalline mass and/or chunks
  • Titration Iodometric:
    ≥97.5 %
  • Sulfated ash:
    ≤0.05 %

 

 

Frequently Bought Together