Order Entry
Italy
ContactUsLinkComponent
 
2,2'-Dipyridyl disulphide for synthesis, Sigma-Aldrich®
Catalog # 8.41109.0005
Supplier:  Merck
CAS Number:  
2,2'-Dipyridyl disulphide for synthesis, Sigma-Aldrich®
Catalog # 8.41109.0005
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Solubility:
    5 g/l (20 °C)
  • Melting Point:
    57 - 61 °C
  • Molar Mass:
    220.3 g/mol
  • Flash Point:
    210 °C
  • Ignition Point:
    340 °C

Specification Test Results

About this item

 :
 :220.32 g/mol
Boiling Pt:348 °C (1013 hPa)
Melting Pt:57…61 °C
 :M-Clarity™ quality level = MQ200
Storage Temperature:Refrigerator
EINECS:218-343-1
CAS Number:2127-03-9
Formula:C₁₀H₈N₂S₂
MDL Number:MFCD00006287