

Dimethyl trisulphide 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3658-80-8
:
Formula:C₂H₆S₃
MDL Number:MFCD00039808
:126.27 g/mol
Melting Pt:–68 °C
Flash Pt:52 °C
EINECS:222-910-9
UN:1993
ADR:3,III
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5995 to 1.6025 (20°C, 589 nm)
- GC:≥98.0 %
Specification Test Results
Frequently Bought Together