Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Selenium dioxide ≥99.4% (metals basis)

Toxic
Health
Environment
Selenium dioxide ≥99.4% (metals basis)
Certificates
Selenium dioxide ≥99.4% (metals basis)
Selenium dioxide ≥99.4% (metals basis)

3 Options Available Below

Product Details & Documents

Purity: 99.4% CAS No.: 7446-08-4 Molecular Formula: O2Se Formula Weight: 110.96 Physical Form: Powder Density: 3.95 Appearance: White to pink Odour: Odourless Melting Point: 340-350° subl. Solubility: Partially soluble in water, methanol, ethanol, acetone. Soluble in concentrated H2SO4 Merck Reference: 14,8434 UN No.: 3283 MDL No.: MFCD00003562

As an oxidizing agent, in manufacturing of other selenium compounds

  • CAS Number:7446-08-4
  • Formula:SeO₂
  • MDL Number:MFCD00003562
  • Molecular Weight:110.96 g/mol
  • Merck Index:14,8434
  • Melting Pt:340…350 °C (subl.)
  • Density:3.95
  • Storage Temperature:Ambient
  • EINECS:231-194-7
  • UN:3283
  • ADR:6.1,II
Product Family Options

Product Information