Order Entry
Italy
ContactUsLinkComponent
Ammonium thiocyanate, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Ammonium thiocyanate, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates
Ammonium thiocyanate, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    1600 g/l (20 °C)
  • Melting Point:
    150 °C
  • Molar Mass:
    76.12 g/mol
  • Bulk Density:
    600 - 700 kg/m3
  • Vapor Pressure:
     - 1 hPa (20 °C)
  • Density:
    1.3 g/cm3 (20 °C)
  • pH:
    4.8 - 5.8 (50 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together