
Ammonium carbonate
Catalog # 4.80014.9025
Supplier: Merck
CAS Number:
Specifications
- Pk:25 L
- Seq:5327
Specifications
About this item
:
:96.09 g/mol
Merck Index:13,00508
Melting Pt:58 °C
Density:1.5 g/cm³ (20 °C)
EINECS:208-058-0
:GHS07
Storage Temperature:Ambient
CAS Number:506-87-6
:10361-29-2
Formula:(NH₄)₂CO₃
MDL Number:MFCD00010890