Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ammonium carbonate
Ammonium carbonate ≥30% (NH₃ basis) ACS
Ammonium carbonate ≥30% (NH₃ basis) ACS
:
Thermo Fisher Scientific
Ammonium carbonate ≥30% (NH₃ basis) ACS
:
Thermo Fisher Scientific
:
506-87-6
:
10361-29-2
:
(NH₄)₂CO₃
:
MFCD00010890
:
96.09 g/mol
:
14,508
:
60 °C (subl.)
:
1.5 g/cm³ (20 °C)
:
208-058-0
:
GHS07
:
Ambient
msds_ALFA036229
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
Zinc sulphate 0.1 mol/l (0.1 N) in aqueous solution, Titripur® Reag. Ph. Eur. volumetric solution, Supelco®
: Merck
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Potassium chloride 99.5-101.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol eurodenatured ≥96%, TechniSolv® (1:1:1)
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Potassium dihydrogen phosphate, EMSURE® ISO for analysis, Supelco®
: Merck
Water USP, JP, Ph. Eur., purified
: VWR Chemicals
Potassium iodide ≥99.5%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Sodium nitrate, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Sodium chloride, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
Sodium dihydrogen phosphate monohydrate 90.0-102.0% (by alkalimetry), EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
VWR® Ready to Use Media Plates (90 mm), Irradiated
: VWR Chemicals