
Tetrapentylammonium bromide 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:866-97-7
:
Formula:C₂₀H₄₄BrN
MDL Number:MFCD00011856
:378.48 g/mol
Melting Pt:100…102 °C
Density:1,0662 g/cm³ (25 °C)
EINECS:212-756-0
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Powder or crystals or flakes
- Infrared spectrum:Conforms
- Melting point:99 to 103 °C
- Titration Argentometric:≥99.0 %
Specification Test Results
Frequently Bought Together